Learn More
Importin alpha 3/KPNA4 Antibody - BSA Free, Novus Biologicals™
Código de producto. 18536074 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.025 mg
0.1 mg
Tamaño de la unidad:
0.10 miligramo
25 microgramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
18536074 0.025 mg 25 microgramos
18525463 0.1 mg 0.10 miligramo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 18536074 Proveedor Novus Biologicals N.º de proveedor NBP1763000.025mg
Rabbit Polyclonal Antibody
Importin alpha 3/KPNA4 Polyclonal antibody specifically detects Importin alpha 3/KPNA4 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Importin alpha 3/KPNA4
Aplicaciones Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Clasificación Polyclonal
Concentración 1 mg/mL
Conjugado Unconjugated
Dilución Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/ Immunofluorescence 2.5-20 μg/mL
Formulación PBS
N.º de referencia del gen EAW78632
Alias de gen importin alpha 3, Importin alpha Q1, importin subunit alpha-4, importin-alpha-Q1, IPOA3, karyopherin alpha 4 (importin alpha 3), MGC12217, MGC26703, Qip1, QIP1Karyopherin subunit alpha-4, SRP3
Especie del huésped Rabbit
Inmunógeno Antibody was raised against a 15 amino acid synthetic peptide near the amino terminus of human KPNA4. The immunogen is located within amino acids 30 - 80 of KPNA4. Amino Acid Squence: RNVPQEDICEDS
Método de purificación Peptide affinity purified
Cantidad 0.025 mg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 3840
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgG