missing translation for 'onlineSavingsMsg'
Learn More

IL-34 Antibody, Alexa Fluor™ 700, R&D Systems™

Código de producto. 18498357 Tienda Bio Techne Productos
Cambiar vista
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad Tamaño de la unidad
18498357 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18498357 Proveedor Novus Biologicals N.º de proveedor NBP176356AF700
 Artículo restringido
Fecha estimada de envoi: 09-10-2026
Añadir a la cesta
Añadir a la cesta

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

IL-34 Polyclonal antibody specifically detects IL-34 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antígeno IL-34
Aplicaciones Western Blot, ELISA
Clasificación Polyclonal
Conjugado Alexa Fluor 700
Formulación 50mM Sodium Borate
Alias de gen C16orf77, IL-34, interleukin 34
Especie del huésped Rabbit
Inmunógeno Antibody was raised against a 13 amino acid synthetic peptide from near the center of human IL-34. The immunogen is located within amino acids 60 - 110 of IL-34. Amino Acid Squence: TRLRAQVSERELR
Método de purificación Peptide affinity purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Signal Transduction
Primario o secundario Primary
ID de gen (Entrez) 146433
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at 4°C in the dark.
Tipo de producto Antibody
Formulario Purified
Isotype IgG
Línea de productos Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.