missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
IL-22R alpha 1 Polyclonal antibody specifically detects IL-22R alpha 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Especificaciones
Especificaciones
| Antígeno | IL-22R alpha 1 |
| Aplicaciones | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Clasificación | Polyclonal |
| Conjugado | DyLight 350 |
| Formulación | 50mM Sodium Borate |
| Alias de gen | CRF2-9interleukin 22 receptor, Cytokine receptor class-II member 9, Cytokine receptor family 2 member 9, IL-22 receptor subunit alpha-1, IL22R, IL22R1, IL-22RA1, IL-22R-alpha-1, interleukin 22 receptor, alpha 1, interleukin-22 receptor subunit alpha-1, zcytoR11 |
| Especie del huésped | Rabbit |
| Inmunógeno | Antibody was raised against a synthetic peptide corresponding to amino acids near the amino terminus of human IL-22 receptor precursor. The immunogen is located within the first 50 amino acids of IL-22 Receptor. Amino Acid Squence: QSSNFENILTWDSGPE |
| Método de purificación | Peptide affinity purified |
| Cantidad | 0.1 mL |
| Visa mer |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?