missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
IgE Monoclonal antibody specifically detects IgE in Human samples. It is validated for Flow Cytometry, ELISA
Specifications
Specifications
| Antígeno | IgE |
| Aplicaciones | Flow Cytometry, ELISA |
| Clasificación | Monoclonal |
| Clon | BE5 |
| Conjugado | DyLight 594 |
| Formulación | 50mM Sodium Borate |
| Alias de gen | IgE, immunoglobulin heavy constant epsilon |
| Especie del huésped | Mouse |
| Inmunógeno | Purified human IgE. |
| Método de purificación | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?