missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human WDR21C (aa 94-118) Control Fragment Recombinant Protein

Código de producto. 30203553
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
30203553 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 30203553 Proveedor Invitrogen™ N.º de proveedor RP103657

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (52%), Rat (52%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-64329 (PA5-64329. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

TRUSTED_SUSTAINABILITY

Specifications

Número de acceso Q8NA75
Concentración ≥5.0 mg/mL
Para utilizar con (aplicación) Blocking Assay, Control
Formulación 1 M urea, PBS with no preservative; pH 7.4
ID de gen (Entrez) 138009
Nombre Human WDR21C (aa 94-118) Control Fragment
Cantidad 100 μL
Estado normativo RUO
Alias de gen DCAF4L2; DDB1 and CUL4 associated factor 4 like 2; DDB1 and CUL4 associated factor 4-like 2; DDB1- and CUL4-associated factor 4-like protein 2; WD repeat domain 21 C; WD repeat-containing protein 21 C; WDR21C
Nombre común WDR21C
Símbolo de gen DCAF4L2
Conjugado Unconjugated
Especie Human
Recombinante Recombinant
Etiqueta de proteína His-ABP-tag
Secuencia AGGSKYGIITMRGLTTPELRVYPHK
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Sistema de expresión E. coli
Formulario Liquid
Grado de pureza o calidad >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.