missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human UBE2E1 (aa 44-146) Control Fragment Recombinant Protein

Código de producto. 30209283
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
30209283 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 30209283 Proveedor Invitrogen™ N.º de proveedor RP102081

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-56476 (PA5-56476. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

UBE2E1 catalyzes the covalent attachment of ubiquitin to other proteins. UBE2E1 mediates the selective degradation of short-lived and abnormal proteins.The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Especificaciones

Número de acceso P51965
Concentración ≥5.0 mg/mL
Para utilizar con (aplicación) Blocking Assay, Control
Formulación 1 M urea, PBS with no preservative; pH 7.4
ID de gen (Entrez) 7324
Nombre Human UBE2E1 (aa 44-146) Control Fragment
Cantidad 100 μL
Estado normativo RUO
Alias de gen (E3-independent) E2 ubiquitin-conjugating enzyme E1; E2 ubiquitin-conjugating enzyme E1; Ubce5; UBCH6; ubcM2; UbcM3; UBE2E1; Ubiquitin carrier protein E1; ubiquitin conjugating enzyme E2 E1; ubiquitin conjugating enzyme E2E 1; ubiquitin-conjugating enzyme 5; ubiquitin-conjugating enzyme E2 E1; Ubiquitin-conjugating enzyme E2 E1-like protein; ubiquitin-conjugating enzyme E2E 1; ubiquitin-conjugating enzyme E2E 1 (homologous to yeast UBC4/5); ubiquitin-conjugating enzyme E2E 1 (UBC4/5 homolog, yeast); ubiquitin-conjugating enzyme E2E 1, UBC4/5 homolog; ubiquitin-protein ligase E1; zgc:171646
Nombre común UBE2E1
Símbolo de gen UBE2E1
Conjugado Unconjugated
Especie Human
Recombinante Recombinant
Etiqueta de proteína His-ABP-tag
Secuencia LLSTSAKRIQKELADITLDPPPNCSAGPKGDNIYEWRSTILGPPGSVYEGGVFFLDITFTPEYPFKPPKVTFRTRIYHCNINSQGVICLDILKDNWSPALTIS
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Sistema de expresión E. coli
Formulario Liquid
Grado de pureza o calidad >80% by SDS-PAGE and Coomassie blue staining
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.