Learn More
Abnova™ Human SLC27A2 Full-length ORF (AAH57770.1, 1 a.a. - 567 a.a.) Recombinant Protein with GST-tag at N-terminal
Description
The protein encoded by this gene is an isozyme of long-chain fatty-acid-coenzyme A ligase family. Although differing in substrate specificity, subcellular localization, and tissue distribution, all isozymes of this family convert free long-chain fatty acids into fatty acyl-CoA esters, and thereby play a key role in lipid biosynthesis and fatty acid degradation. This isozyme activates long-chain, branched-chain and very-long-chain fatty acids containing 22 or more carbons to their CoA derivatives. It is expressed primarily in liver and kidney, and is present in both endoplasmic reticulum and peroxisomes, but not in mitochondria. Its decreased peroxisomal enzyme activity is in part responsible for the biochemical pathology in X-linked adrenoleukodystrophy. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Specifications
Specifications
| Número de acceso | AAH57770.1 |
| Para utilizar con (aplicación) | Antibody Production, Protein Array, ELISA, Western Blot |
| Formulación | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
| ID de gen (Entrez) | 11001 |
| Peso molecular | 91kDa |
| Nombre | SLC27A2 (Human) Recombinant Protein (P01) |
| Método de purificación | Glutathione Sepharose 4 Fast Flow |
| Pruebas de control de calidad | 12.5% SDS-PAGE Stained with Coomassie Blue. |
| Cantidad | 2 μg |
| Inmunógeno | MLSAIYTVLAGLLFLPLLVNLCCPYFFQDIGYFLKVAAVGRRVRSYGKRRPARTILRAFLEKARQTPHKPFLLFRDETLTYAQVDRRSNQVARALHDHLGLRQGDCVALLMGNEPAYVWLWLGLVKLGCAMACLNYNIRAKSLLHCFQCCGAKVLLVSPELQAAVEEILPSLKKDDVSIYYVSRTSNTDGIDSFLDKVDEVSTEPIPESWRSEVTFSTPALYIYTSGTTGATLALRTKFSASQFWDDCRKYNVTVIQYIGELLRYLCNSPQKPNDRDHKVRLALGNGLRGDVWRQFVKRFGDICIYEFYAATEGNIGFMNYARKVGAVGRVNYLQKKIITYDLIKYDVEKDEPVRDENGYCVRVPKGEVGLLVCKITQLTPFNGYAGAKAQTEKKKLRDVFKKGDLYFNSGDLLMVDHENFIYFHDRVGDTFRWKGENVATTEVADTVGLVDFVQEVNVYGVHVPDHEGRIGMASIKMKENHEFDGKKLFQHIADYLPSYARPRFLRIQDTIEITGTFKHRKMTLVEEGFNPAVIKDALYFLDDTAKMYVPMTEDIYNAISAKTLKL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.