Learn More
Invitrogen™ Human SLC16A9 (aa 191-255) Control Fragment Recombinant Protein
Código de producto. 30194064
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
30194064 100 μL 100 microlitros
1 options
Código de producto. 30194064 Proveedor Invitrogen™ N.º de proveedor RP97135
Recombinant Protein
Highest antigen sequence indentity to the following orthologs: Mouse (60%), Rat (60%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61924 (PA5-61924. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Predicted to enable monocarboxylic acid transmembrane transporter activity. Involved in urate metabolic process. Predicted to be integral component of plasma membrane.
TRUSTED_SUSTAINABILITY
Especificaciones
Número de acceso Q7RTY1
Concentración ≥5.0 mg/mL
Para utilizar con (aplicación) Blocking Assay, Control
Formulación 1 M urea, PBS with no preservative; pH 7.4
ID de gen (Entrez) 220963
Nombre Human SLC16A9 (aa 191-255) Control Fragment
Cantidad 100 μL
Estado normativo RUO
Alias de gen 1200003C15Rik; 4930425B13Rik; AI552636; C10orf36; I79_016926; MCT 9; MCT9; monocarboxylate transporter; Monocarboxylate transporter 9; monocarboxylic acid transporter 9; Slc16a9; solute carrier family 16 (monocarboxylic acid transporters), member 9; solute carrier family 16 member 9; solute carrier family 16, member 9; solute carrier family 16, member 9 (monocarboxylic acid transporter 9)
Nombre común SLC16A9
Símbolo de gen SLC16A9
Conjugado Unconjugated
Especie Human
Recombinante Recombinant
Etiqueta de proteína His-ABP-tag
Secuencia LQSSDCPLPKKIAPEDLPDKYSIYNEKGKNLEENINILDKSYSSEEKCRITLANGDWKQDSLLHK
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Sistema de expresión E. coli
Formulario Liquid
Grado de pureza o calidad >80% by SDS-PAGE and Coomassie blue staining