missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SHPRH Partial ORF (NP_775105, 1560 a.a. - 1659 a.a.) Recombinant Protein with GST-tag at N-terminal
Descripción
SHPRH is a ubiquitously expressed protein that contains motifs characteristics of several DNA repair proteins, transcription factors, and helicases. SHPRH is a functional homolog of S. cerevisiae RAD5 (Unk et al., 2006 [PubMed 17108083]).[supplied by OMIM]
Sequence: AQISRVKTFQENLSAFKRDPQINILLLPLHTGSNGLTIIEATHVLLVEPILNPAHELQAIGRVHRIGQTKPTIVHRFLIKATIEERMQAMLKTAERSIYI
Especificaciones
Especificaciones
| Número de acceso | NP_775105 |
| Para utilizar con (aplicación) | Antibody Production, ELISA, Protein Array, Western Blot |
| Formulación | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
| ID de gen (Entrez) | 257218 |
| Peso molecular | 36.74kDa |
| Nombre | SHPRH (Human) Recombinant Protein (Q01) |
| Pruebas de control de calidad | 12.5% SDS-PAGE Stained with Coomassie Blue. |
| Cantidad | 25 μg |
| Inmunógeno | AQISRVKTFQENLSAFKRDPQINILLLPLHTGSNGLTIIEATHVLLVEPILNPAHELQAIGRVHRIGQTKPTIVHRFLIKATIEERMQAMLKTAERSIYI |
| Requisitos de almacenamiento | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Mostrar más |
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?