missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human NFAT5 (aa 1106-1190) Control Fragment Recombinant Protein

Código de producto. 30210675
Cambiar vista
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
30210675 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 30210675 Proveedor Invitrogen™ N.º de proveedor RP109813

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (84%), Rat (84%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-145064 (PA5-145064. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The nuclear factor of activated T-cells (NFAT) transcription complex is required for the expression of a group of proteins that collectively regulate the immune response. Four NFAT proteins, encoded on separate genes and expressed as several splice variants, have been described: NFAT1 (also known as NFATp or NFATc2), NFAT2 (NFATc or NFATc1), NFAT3, and NFAT4 (NFATx or NFATc3). These proteins show a low level of sequence similarity with the Dorsal/Rel/NFkB family of transcription factors. Another NFAT-related protein termed NFAT5 differs from isoforms 1-4 in that it lacks many of the Fos/Jun contact sites observed in its predecessors and its subcellular localization is not calcineurin-dependent.
TRUSTED_SUSTAINABILITY

Especificaciones

Número de acceso O94916
Concentración ≥5.0 mg/mL
Para utilizar con (aplicación) Blocking Assay, Control
Formulación 1 M urea, PBS with no preservative; pH 7.4
ID de gen (Entrez) 10725
Nombre Human NFAT5 (aa 1106-1190) Control Fragment
Cantidad 100 μL
Estado normativo RUO
Alias de gen AI225870; B130038B15Rik; CAG-8; CAG80; glutamine rich protein H65; KIAA0827; mKIAA0827; Nfat; Nfat5; NF-AT5; NFATL1; NFAT-like protein 1; nfatz; nuclear factor of activated T cells 5; nuclear factor of activated T-cells 5; nuclear factor of activated T-cells 5, tonicity-responsive; OREBP; osmotic response element-binding protein; rel domain-containing transcription factor NFAT5; T-cell transcription factor NFAT5; TonE-binding protein; TONEBP; tonicity-responsive enhancer binding protein; tonicity-responsive enhancer-binding protein
Nombre común NFAT5
Símbolo de gen NFAT5
Conjugado Unconjugated
Especie Human
Recombinante Recombinant
Etiqueta de proteína His-ABP-tag
Secuencia TTSSEQMQPPMFHSQSTIAVLQGSSVPQDQQSTNIFLSQSPMNNLQTNTVAQEAFFAAPNSISPLQSTSNSEQQAAFQQQAPISH
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Sistema de expresión E. coli
Formulario Liquid
Grado de pureza o calidad >80% by SDS-PAGE and Coomassie blue staining
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.