missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human IL-27 Recombinant Protein, PeproTech®

Código de producto. 17831613
Change view
Click to view available options
Cantidad:
100 μg
10 μg
1 mg
250 μg
2 μg
500 μg
50 μg
Tamaño de la unidad:
1 miligramo
10 microgramos
100 microgramos
2 microgramos
250 microgramos
50 microgramos
500 microgramos
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
17831613 2 μg 2 microgramos
17820643 100 μg 100 microgramos
17810643 10 μg 10 microgramos
17821213 1 mg 1 miligramo
17830083 250 μg 250 microgramos
17890913 500 μg 500 microgramos
17850063 50 μg 50 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 17831613 Proveedor Gibco™ N.º de proveedor 200382UG

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Protein

Recombinant Human IL-27 produced from HEK293 cells is a non-disulfide-linked, heterodimeric protein composed of a 209 amino acid length EBI3 subunit and a 215 amino acid length IL-27-p28 subunit, for a total sequence length of 424 amino acid residues. The calculated molecular weight of the associated IL-27 subunits is 47.8 kDa. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

IL-27 (Interleukin 27) is a cytokine belonging to the IL-12 family, and is a heterodimeric cytokine composed of EBI3 and p28 subunits. IL-27 is produced by activated dendritic cells and macrophages in response to Toll-like receptor ligands and pro-inflammatory cytokines. IL-27 promotes CD4+ T cell differentiation to the Th1 lineage by inducing expression of the transcription factor T-bet and up regulating IL-12R beta2, which in turn suppresses Th2 and Th17 differentiation and proliferation. IL-27 is a recently discovered member of the IL-6/IL-12 family of proinflammatory and immunoregulatory cytokines. IL-27 acts by binding to its receptor WSX-1 and gp130 which results in the activation of a Jak/STAT signaling cascade, suggesting IL-27 involvement in the regulation of immune processes. It has been suggested that IL-27 can also be used as a therapeutic agent against cancer as it can also induce tumor-specific anti-tumor activity mediated through CD8+ T-cells, IFN-gamma, and T-bet. In humans, the gene is located on the p arm of chromosome 16.
TRUSTED_SUSTAINABILITY

Especificaciones

Número de acceso Q14213, Q8NEV9
Para utilizar con (aplicación) Control, Bioactivity
Formulación Protein with no preservative
ID de gen (Entrez) 10148, 246778
Peso molecular 47.8 kDa
Nombre Human IL-27
Método de purificación Purified
Cantidad 2 μg
Fuente HEK293 cells
Requisitos de almacenamiento -20°C
Estado normativo RUO
Concentración de endotoxinas <1 EU/μg
Alias de gen cytokine receptor; EBI3; EBI-3; EBV-induced gene 3 protein; Epstein-Barr virus induced 3; Epstein-Barr virus induced gene 3; Epstein-Barr virus induced gene 3 (similar to Interleukin-27 beta chain precursor); epstein-Barr virus-induced gene 3 protein; Epstein-Barr virus-induced gene 3 protein homolog; Il27; IL-27; IL27 p28; IL-27 p28 subunit; IL27 subunit; IL-27 subunit alpha; IL-27 subunit beta; Il27a; IL-27A; IL-27-A; IL27-A; Il27b; IL-27B; IL27p28; IL-27p28; Il30; IL35 subunit; ILN; Interleukin; interleukin 27; interleukin 30; interleukin-27 subunit alpha; Interleukin-27 subunit beta; interleukin-30; MGC71873; M-IL-27 (p28); p28; RGD1561420
Nombre común IL-27
Símbolo de gen Ebi3, IL27
Actividad biológica Determined by its ability to stimulate the proliferation of human TF-1 cells. The expected ED50 range is 400-500 ng/ml
Tipo de producto Protein
Conjugado Unconjugated
Recombinante Recombinant
Secuencia p28 subunit: FPRPPGRPQL SLQELRREFT VSLHLARKLL SEVRGQAHRF AESHLPGVNL YLLPLGEQLP DVSLTFQAWR RLSDPERLCF ISTTLQPFHA LLGGLGTQGR WTNMERMQLW AMRLDLRDLQ RHLRFQVLAA GFNLPEEEEE EEEEEEEERK GLLPGALGSA LQGPAQVSWP QLLSTYRLLH SLELVLSRAV RELLLLSKAG HSVWPLGFPT LSPQP EBI3 subunit: RKGPPAALTL PRVQCRASRY PIAVDCSWTL PPAPNSTSPV SFIATYRLGM AARGHSWPCL QQTPTSTSCT ITDVQLFSMA PYVLNVTAVH PWGSSSSFVP FITEHIIKPD PPEGVRLSPL AERQLQVQWE PPGSWPFPEI FSLKYWIRYK RQGAARFHRV GPIEATSFIL RAVRPRARYY VQVAAQDLTD YGELSDWSLP ATATMSLGK
Contenido y almacenamiento -20°C
Actividad Determined by its ability to stimulate the proliferation of human TF-1 cells. The expected ED50 range is 400-500 ng/ml.
Sistema de expresión HEK293 cells
Formulario Lyophilized
Grado de pureza o calidad ≥ 90% by SDS-PAGE gel and HPLC analyses.
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.