missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human GDF-5, Animal-Free Recombinant Protein, PeproTech®

Código de producto. 17806748
Change view
Click to view available options
Cantidad:
250 μg
50 μg
Tamaño de la unidad:
250 microgramos
50 microgramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
17806748 250 μg 250 microgramos
17866448 50 μg 50 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 17806748 Proveedor Gibco™ N.º de proveedor AF12001250UG

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Protein

Recombinant Human GDF-5 (BMP-14/CDMP-1) is a 27.0 kDa homodimeric disulfide-linked protein consisting of two 120 amino acid polypeptide chains. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

GDF-5 is expressed in long bones during embryonic development and postnatally in articular cartilage. Mutations in the GDF-5 gene have been implicated in Hunter-Thompson type dwarfism and in Grebe Syndrome, which is characterized by short stature, extra digits, and short and deformed extremities. The mature and functional form of GDF-5 is a homodimer of two 120 amino-acid polypeptide chain (monomers) linked by a single disulfide bond. Each GDF-5 monomer is expressed as the C-terminal part of a precursor polypeptide, which also contains a 27 amino acid signal peptide and a 354 amino acid propeptide. This precursor undergoes intracellular dimerization, and upon secretion it is processed by a furin-type protease.
TRUSTED_SUSTAINABILITY

Especificaciones

Número de acceso P43026
Para utilizar con (aplicación) Functional Assay
Formato Lyophilized
Formulación protein with no preservative
ID de gen (Entrez) 8200
Peso molecular 27 kDa
Nombre Human GDF-5, Animal-Free
Método de purificación HPLC, SDS-PAGE
Cantidad 250 μg
Estado normativo RUO
Concentración de endotoxinas <0.1 EU/ μg
Alias de gen BDA1C; Bmp14; BMP-14; Bone morphogenetic protein 14; bp; brachypodism; brp; Cartilage-derived morphogenetic protein 1; cartilage-derived morphogenetic protein-1; CDMP1; Cdmp-1; GDF5; GDF-5; growth differentiation factor 5; growth/differentiation factor 5; H-GDF-5; LAP4; LAP-4; Lipopolysaccharide-associated protein 4; LPS-associated protein 4; OS5; radotermin; SYM1B; SYNS2
Nombre común GDF5
Símbolo de gen GDF5
Actividad biológica Determined by its ability to induce alkaline phosphatase production by ATDC-5 cells. The expected ED50 for this effect is 1.0-2.0 ug /ml.
Tipo de producto Protein
Conjugado Unconjugated
Recombinante Recombinant
Secuencia APLATRQGKR PSKNLKARCS RKALHVNFKD MGWDDWIIAP LEYEAFHCEG LCEFPLRSHL EPTNHAVIQT LMNSMDPEST PPTCCVPTRL SPISILFIDS ANNVVYKQYE DMVVESCGCR
Contenido y almacenamiento -20°C
Actividad Determined by its ability to induce alkaline phosphatase production by ATDC-5 cells. The expected ED50 for this effect is 1.0-2.0 ug /ml.
Sistema de expresión E. coli
Formulario Lyophilized
Grado de pureza o calidad ≥ 98% by SDS-PAGE gel and HPLC analyses.
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.