missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ETV3 (aa 399-499) Control Fragment Recombinant Protein

Código de producto. 30212344
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
30212344 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 30212344 Proveedor Invitrogen™ N.º de proveedor RP102505

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (76%), Rat (76%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-52107 (PA5-52107. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Transcriptional repressor that contribute to growth arrest during terminal macrophage differentiation by repressing target genes involved in Ras-dependent proliferation. Represses MMP1 promoter activity.
TRUSTED_SUSTAINABILITY

Especificaciones

Número de acceso P41162
Concentración ≥5.0 mg/mL
Para utilizar con (aplicación) Blocking Assay, Control
Formulación 1 M urea, PBS with no preservative; pH 7.4
ID de gen (Entrez) 2117
Nombre Human ETV3 (aa 399-499) Control Fragment
Cantidad 100 μL
Estado normativo RUO
Alias de gen AI414410; bA110J1.4; ETS domain transcriptional repressor PE1; ETS translocation variant 3; ETS variant 3; ets variant gene 3; ets variant gene 3, ETS family transcriptional repressor; ETS-domain protein; ETS-domain transcriptional repressor; Etv3; Mets; Mitogenic Ets transcriptional suppressor; mitogenic Ets transcriptional suppressor METS; Pe1; PE-1; SH2 domain containing phosphatase anchor protein 1; Spap1
Nombre común ETV3
Símbolo de gen ETV3
Conjugado Unconjugated
Especie Human
Recombinante Recombinant
Etiqueta de proteína His-ABP-tag
Secuencia EKEEHTQEEGTVPSRTIEEEKGTIFARPAAPPIWPSVPISTPSGEPLEVTEDSEDRPGKEPSAPEKKEDALMPPKLRLKRRWNDDPEARELSKSGKFLWNG
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Sistema de expresión E. coli
Formulario Liquid
Grado de pureza o calidad >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.