missing translation for 'onlineSavingsMsg'
Learn More

HPV16 Antibody (rHPV16L1/1058), Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18591724 Shop All Bio Techne Products
Change view
Click to view available options
Cantidad:
100 μg
Unit Size:
100 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
18591724 100 μg 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18591724 Supplier Novus Biologicals Supplier No. NBP308318JF549

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HPV16 Monoclonal antibody specifically detects HPV16 in Virus samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antígeno HPV16
Aplicaciones Immunohistochemistry (Paraffin)
Clasificación Monoclonal
Clon rHPV16L1/1058
Conjugado Janelia Fluor 549
Formulación 50mM Sodium Borate
Alias de gen Human Papilloma Virus Type 16
Especie del huésped Mouse
Inmunógeno Human HPV16, major capsid protein L1
Método de purificación Protein A or G purified
Cantidad 100 μg
Estado normativo RUO
Disciplina de investigación Virology Bacteria and Parasites
Primario o secundario Primary
Especies diana Virus
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.