missing translation for 'onlineSavingsMsg'
Learn More

Host cell factor C2 Antibody [DyLight 650], Novus Biologicals™

Código de producto. 30566431 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
30566431 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 30566431 Proveedor Novus Biologicals N.º de proveedor NBP346141C

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Host cell factor C2 Polyclonal antibody specifically detects Host cell factor C2 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antígeno Host cell factor C2
Aplicaciones ELISA, Western Blot
Clasificación Polyclonal
Conjugado DyLight 650
Formulación 50mM Sodium Borate
Alias de gen FLJ94012, HCF2, HCF-2C2 factor, host cell factor 2, host cell factor C2
Especie del huésped Rabbit
Inmunógeno host cell factor C2. (Uniprot# Q9Y5Z7)
Método de purificación Immunogen affinity purified
Cantidad 0.1 mL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 29915
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.