missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human GRK4, GST Tag Recombinant Protein

Código de producto. 15438543
Cambiar vista
Click to view available options
Cantidad:
100 μg
1 mg
Tamaño de la unidad:
1 miligramo
100 microgramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
15438543 100 μg 100 microgramos
10706023 1 mg 1 miligramo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 15438543 Proveedor Invitrogen™ N.º de proveedor PR7249A

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length

GRK4 is a member of the G protein-coupled receptor kinase subfamily of the serine/threonine protein kinase family. GRK4 phosphorylates the activated forms of G protein-coupled receptors. GRK4 has been linked to both genetic and acquired hypertension.
TRUSTED_SUSTAINABILITY

Especificaciones

Número de acceso P32298
Concentración See Label
Para utilizar con (aplicación) Kinase Assay
Formulación 50 mM tris with 0.02% Triton X-100, 0.5 mM EDTA, 150 mM NaCl, 2 mM DTT, 50% glycerol and no preservative; pH 7.5
ID de gen (Entrez) 2868
Peso molecular 94 kDa
Nombre Human GRK4, GST Tag
Cantidad 100 μg
Estado normativo RUO - research use only
Alias de gen G protein-coupled receptor kinase 4; G protein-coupled receptor kinase GRK4; GPRK2L; GPRK4; GRK4; GRK4a; IT11; ITI1
Nombre común GRK4
Familia Quinasa GRK Kinase Family
Conjugado Unconjugated
Especie Human
Recombinante Recombinant
Etiqueta de proteína GST-tag
Secuencia Full length
Contenido y almacenamiento -80°C, Avoid Freeze/Thaw Cycles
Sistema de expresión Insect cells
Formulario Liquid
Mostrar más Mostrar menos
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.