Learn More
Goat anti-Human Kappa Light Chain Secondary Antibody, DyLight 680, Novus Biologicals™
Tienda Bio Techne ProductosDescripción
Kappa Light Chain Polyclonal antibody specifically detects Kappa Light Chain in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunodiffusion, Immunoassay
Especificaciones
Especificaciones
| Antígeno | Kappa Light Chain |
| Aplicaciones | Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunodiffusion |
| Clasificación | Polyclonal |
| Conjugado | DyLight 680 |
| Dilución | Western Blot : 1:100-1:2000, ELISA : 1:100-1:2000, Immunohistochemistry : 1:10 - 1:500, Immunoprecipitation : 1:10 - 1:500, Immunodiffusion, Immunoassay |
| Formulación | 50mM Sodium Borate |
| Alias de gen | HCAK1, IGKCD, immunoglobulin kappa constant, Km, MGC111575, MGC62011, MGC72072, MGC88770, MGC88771, MGC88809 |
| Especie del huésped | Goat |
| Inmunógeno | Purified Human Kappa Chain |
| Método de purificación | Affinity purified |
| Mostrar más |