Learn More
GNE-617 hydrochloride, MedChemExpress
Código de producto. 16492877
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
2 mg
5 mg
10 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
2 miligramos
5 miligramos
50 miligramos
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
16492877 50 mg 50 miligramos
16480256 2 mg 2 miligramos
16476808 10 mg 10 miligramos
16429357 5 mg 5 miligramos
16426926 100 mg 100 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Código de producto. 16492877 Proveedor Medchem Express N.º de proveedor HY15766A
MedChemExpress GNE-617 hydrochloride is a specific NAMPT inhibitor that inhibits the biochemical activity of NAMPT with an IC50 of 5 nM and exhibits efficacy in xenograft models of cancer.
GNE-617 hydrochloride is a specific NAMPT inhibitor that inhibits the biochemical activity of NAMPT with an IC50 of 5 nM and exhibits efficacy in xenograft models of cancer.
Especificaciones
Nombre del producto químico o material GNE-617 hydrochloride
CAS 2070014-99-0
Color Off-White
Notas de grado de pureza Research
Fórmula molecular C21H16ClF2N3O3S
Cantidad 50 mg
Información de solubilidad DMSO : 5.56 mg/mL (11.99 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(NCC1=CC=C(S(C2=CC(F)=CC(F)=C2)(=O)=O)C=C1)C3=CN4C(C=C3)=NC=C4.Cl
Peso molecular (g/mol) 463.88
Formula Weight (peso de la fórmula) 463.88
Porcentaje de pureza 99.15%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Cancer-programmed cell death