missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
Glut1 Polyclonal antibody specifically detects Glut1 in Human, Mouse, Rat, Rabbit samples. It is validated for Western Blot, ChIP assay, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), In vitro Assay, Flow (Intracellular), ChIP assay (ChIP)
Especificaciones
Especificaciones
| Antígeno | Glut1 |
| Aplicaciones | Western Blot, ChIP Assay, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence |
| Clasificación | Polyclonal |
| Conjugado | Alexa Fluor 647 |
| Formulación | 50 mM Sodium Borate |
| Alias de gen | Choreoathetosis/Spasticity, Episodic (Paroxysmal Choreoathetosis), CSE, DYT17, DYT18, DYT9, EIG12, Glucose transporter type 1, erythrocyte/brain, GLUT-1, GLUT1DS, HepG2 glucose transporter, HTLVR, Human T-Cell Leukemia Virus (I and II) Receptor, MGC141895, MGC141896, PED, solute carrier family 2 (facilitated glucose transporter), member 1, Solute Carrier Family 2 Member 1, solute carrier family 2, facilitated glucose transporter member 1 |
| Especie del huésped | Rabbit |
| Inmunógeno | This Glut1 antibody is made against a synthetic peptide made to an N-terminal region of the human GLUT1 protein (between residues 1-100). [Swiss-Prot# P11166]. The immunogen is cytosolic. |
| Método de purificación | Immunogen affinity purified |
| Cantidad | 0.1 mL |
| Mostrar más |
Título del producto
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?