missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
GLI4 Polyclonal antibody specifically detects GLI4 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Especificaciones
Especificaciones
| Antígeno | GLI4 |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Polyclonal |
| Conjugado | Janelia Fluor 669 |
| Formulación | 50mM Sodium Borate |
| Alias de gen | GLI family zinc finger 4, GLI-Kruppel family member GLI4, glioma-associated oncogene family zinc finger 4, HKR4GLI-Kruppel family member GLI4 (oncogene HKR4), Krueppel-related zinc finger protein 4, Protein HKR4, zinc finger protein GLI4, ZNF928 |
| Especie del huésped | Rabbit |
| Inmunógeno | GLI family zinc finger 4. (Uniprot# P10075) |
| Método de purificación | Immunogen affinity purified |
| Cantidad | 0.1 mL |
| Mostrar más |
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?