missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GDF-5/BMP-14 Antibody (1065032) [mFluor Violet 450 SE], Novus Biologicals™
Tienda Bio Techne ProductosDescripción
GDF-5/BMP-14 Monoclonal antibody specifically detects GDF-5/BMP-14 in Human samples. It is validated for Immunocytochemistry
Especificaciones
Especificaciones
| Antígeno | GDF-5/BMP-14 |
| Aplicaciones | Immunocytochemistry |
| Clasificación | Monoclonal |
| Clon | 1065032 |
| Conjugado | mFluor Violet 450 SE |
| Formulación | 50mM Sodium Borate |
| Alias de gen | BMP14, Cartilage-derived morphogenetic protein 1, cartilage-derived morphogenetic protein-1, CDMP-1, CDMP1LAP4, GDF-5, growth differentiation factor 5, growth/differentiation factor 5, OS5, Radotermin, SYNS2 |
| Especie del huésped | Mouse |
| Inmunógeno | E. coli-derived recombinant human GDF-5, Ala382-Arg501, Accession # P43026 |
| Método de purificación | Protein A or G purified from hybridoma culture supernatant |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?