missing translation for 'onlineSavingsMsg'
Learn More

GDF-5/BMP-14 Antibody (1065032) [DyLight 350], Novus Biologicals™

Código de producto. 30671439 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
30671439 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 30671439 Proveedor Novus Biologicals N.º de proveedor FAB8340D

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

GDF-5/BMP-14 Monoclonal antibody specifically detects GDF-5/BMP-14 in Human samples. It is validated for Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antígeno GDF-5/BMP-14
Aplicaciones Immunocytochemistry
Clasificación Monoclonal
Clon 1065032
Conjugado DyLight 350
Formulación 50mM Sodium Borate
Alias de gen BMP14, Cartilage-derived morphogenetic protein 1, cartilage-derived morphogenetic protein-1, CDMP-1, CDMP1LAP4, GDF-5, growth differentiation factor 5, growth/differentiation factor 5, OS5, Radotermin, SYNS2
Especie del huésped Mouse
Inmunógeno E. coli-derived recombinant human GDF-5, Ala382-Arg501, Accession # P43026
Método de purificación Protein A or G purified from hybridoma culture supernatant
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Cytokine Research
Primario o secundario Primary
ID de gen (Entrez) 8200
Especies diana Human
Contenido y almacenamiento Store at 4C in the dark.
Formulario Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.