missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
GDAP1L1 Polyclonal antibody specifically detects GDAP1L1 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot
Especificaciones
Especificaciones
| Antígeno | GDAP1L1 |
| Aplicaciones | ELISA, Immunohistochemistry, Western Blot |
| Clasificación | Polyclonal |
| Conjugado | APC |
| Formulación | PBS |
| Alias de gen | dJ881L22.1, dJ995J12.1.1, DKFZp761K228, ganglioside-induced differentiation-associated protein 1-like 1, GDAP1-L1 |
| Especie del huésped | Rabbit |
| Inmunógeno | ganglioside-induced differentiation-associated protein 1-like 1. (Uniprot# Q96MZ0) |
| Método de purificación | Immunogen affinity purified |
| Cantidad | 0.1 mL |
| Mostrar más |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?