missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
GAS8 Polyclonal antibody specifically detects GAS8 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Especificaciones
Especificaciones
| Antígeno | GAS8 |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Polyclonal |
| Conjugado | FITC |
| Formulación | PBS |
| Alias de gen | GAS-11, GAS11MGC138326, GAS-8, growth arrest specific 11, growth arrest-specific 11, growth arrest-specific 8, Growth arrest-specific protein 11, growth arrest-specific protein 8 |
| Especie del huésped | Rabbit |
| Inmunógeno | growth arrest-specific 8. (Uniprot# O95995) |
| Método de purificación | Immunogen affinity purified |
| Cantidad | 0.1 mL |
| Mostrar más |
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?