missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
FXYD7 Polyclonal antibody specifically detects FXYD7 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
Specifications
| Antígeno | FXYD7 |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Polyclonal |
| Conjugado | Janelia Fluor 525 |
| Formulación | 50mM Sodium Borate |
| Alias de gen | FLJ25096, FXYD domain containing ion transport regulator 7, FXYD domain-containing ion transport regulator 7 |
| Especie del huésped | Rabbit |
| Inmunógeno | FXYD domain containing ion transport regulator 7. (Uniprot# P58549) |
| Método de purificación | Immunogen affinity purified |
| Cantidad | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?