Learn More
FSHR Antibody (6E8.2F5) - BSA Free, Novus Biologicals™
Código de producto. 18786353 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.025 mg
0.1 mg
Tamaño de la unidad:
0.10 miligramo
25 microgramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
18786353 0.1 mg 0.10 miligramo
18770194 0.025 mg 25 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 18786353 Proveedor Novus Biologicals N.º de proveedor NBP236489
Mouse Monoclonal Antibody has been used in 2 publications
FSHR Monoclonal specifically detects FSHR in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno FSHR
Aplicaciones Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Clasificación Monoclonal
Clon 6E8.2F5
Concentración 1.0 mg/mL
Conjugado Unconjugated
Dilución Western Blot 2-3 ug/ml, Immunohistochemistry 2-5 ug/ml, Immunohistochemistry-Paraffin 2-5 ug/ml
Formulación PBS with 0.05% Sodium Azide
N.º de referencia del gen P23945
Alias de gen follicle stimulating hormone receptor, follicle-stimulating hormone receptor, Follitropin receptor, FSH receptor, FSH-R, LGR1FSHRO, MGC141667, MGC141668, ODG1
Símbolos de los genes FSHR
Especie del huésped Mouse
Inmunógeno A synthetic peptide made to the human FSH R protein sequence (between residues 1-200). [Swiss-Prot P23945]
Método de purificación Protein G purified
Cantidad 0.1 mg
Estado normativo RUO
Disciplina de investigación GPCR
Primario o secundario Primary
ID de gen (Entrez) 2492
Especificidad de la prueba Immunogen's sequence similarity with other species: Chicken and Duck (73%), Xenopus (71%).
Especies diana Human
Contenido y almacenamiento Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Formulario Purified
Mostrar más Mostrar menos
For Research Use Only