missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
FOXS1 Polyclonal antibody specifically detects FOXS1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Especificaciones
Especificaciones
| Antígeno | FOXS1 |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Polyclonal |
| Conjugado | FITC |
| Formulación | PBS |
| Alias de gen | FKHL18, forkhead (Drosophila)-like 18, forkhead box protein S1, forkhead box S1, Forkhead-like 18 protein, forkhead-related activator 10, Forkhead-related transcription factor 10, forkhead-related transcription factor FREAC-10, FREAC-10, FREAC10forkhead-like 18 (Drosophila), MGC4544 |
| Especie del huésped | Rabbit |
| Inmunógeno | forkhead box S1. (Uniprot# O43638) |
| Método de purificación | Immunogen affinity purified |
| Cantidad | 0.1 mL |
| Mostrar más |
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?