missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
FMR1NB Polyclonal antibody specifically detects FMR1NB in Human samples. It is validated for ELISA,Western Blot
Especificaciones
Especificaciones
| Antígeno | FMR1NB |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Polyclonal |
| Conjugado | DyLight 405 |
| Formulación | 50mM Sodium Borate |
| Alias de gen | Cancer/testis antigen 37, CT37Sarcoma antigen NY-SAR-35, FLJ25736, fragile X mental retardation 1 neighbor, fragile X mental retardation 1 neighbor protein, NYSAR35, NY-SAR-35 |
| Especie del huésped | Rabbit |
| Inmunógeno | fragile X mental retardation 1 neighbor. (Uniprot# Q8N0W7) |
| Método de purificación | Immunogen affinity purified |
| Cantidad | 0.1 mL |
| Mostrar más |
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?