Learn More
Filanesib, MedChemExpress
Código de producto. 16498937
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
5 miligramos
50 miligramos
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
16498937 5 mg 5 miligramos
16442467 50 mg 50 miligramos
16406666 100 mg 100 miligramos
16406358 10 mg 10 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Código de producto. 16498937 Proveedor Medchem Express N.º de proveedor HY15187
MedChemExpress Filanesib (ARRY-520) is a selective and noncompetitive kinesin spindle protein (KSP) inhibitor, with an IC50 of 6 nM for human KSP. Filanesib induces cell death by apoptosis in vitro. Filanesib has potent anti-proliferative activity.
Filanesib (ARRY-520) is a selective and noncompetitive kinesin spindle protein (KSP) inhibitor, with an IC50 of 6 nM for human KSP. Filanesib induces cell death by apoptosis in vitro. Filanesib has potent anti-proliferative activity.
Especificaciones
Nombre del producto químico o material Filanesib
CAS 885060-09-3
Color Blanco
Notas de grado de pureza Research
Fórmula molecular C20H22F2N4O2S
Cantidad 5 mg
Sinónimo ARRY-520
Información de solubilidad DMSO : ≥ 100 mg/mL (237.82 mM)
SMILES O=C(N1[C@@](C2=CC=CC=C2)(CCCN)SC(C3=CC(F)=CC=C3F)=N1)N(OC)C
Peso molecular (g/mol) 420.48
Formula Weight (peso de la fórmula) 420.48
Porcentaje de pureza 95.0%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Cancer-Kinase/protease