Learn More
Fc epsilon RI Antibody (9E1) - BSA Free, Novus Biologicals™
Código de producto. 18033646 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.025 mg
0.1 mg
Tamaño de la unidad:
0.10 miligramo
25 microgramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
18033646 0.025 mg 25 microgramos
18096564 0.1 mg 0.10 miligramo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 18033646 Proveedor Novus Biologicals N.º de proveedor NB100632690.025MG
Mouse Monoclonal Antibody
Fc epsilon RI Monoclonal specifically detects Fc epsilon RI in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Fc epsilon RI
Aplicaciones Immunoprecipitation, Flow Cytometry, Immunocytochemistry, Immunofluorescence, ELISA, Western Blot
Clasificación Monoclonal
Clon 90
Concentración 1.0 mg/mL
Conjugado Unconjugated
Dilución Western Blot 1:100-1:2000, Flow Cytometry 1:50-1:100, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunoprecipitation 1:10-1:500
Formulación PBS (pH 7.4) with 0.09% Sodium Azide
N.º de referencia del gen P12319
Alias de gen alpha polypeptide, Fc fragment of IgE, high affinity I, receptor for; alpha polypeptide, Fc-epsilon RI-alpha, FcERI, high affinity immunoglobulin epsilon receptor alpha-subunit, high affinity immunoglobulin epsilon receptor subunit alpha, high-affinity, of mast cells, alpha polypeptide
Símbolos de los genes FCER1A
Especie del huésped Mouse
Inmunógeno Made to Human Fc EPSILON R1 ALPHA
Método de purificación Protein G purified
Cantidad 0.025 mg
Estado normativo RUO
Disciplina de investigación Apoptosis, Asthma, Cancer, Immunology, Signal Transduction
Primario o secundario Primary
ID de gen (Entrez) 2205
Especificidad de la prueba NB100-63269 recognizes the human high affinity Fc receptor for IgE (Fc epsilon R1), which plays a central role in the IgE-mediated allergic response. Fc epsilon R1 consists of four subunits, a high affinity IgE binding alpha subunit, a beta chain and two di-sulphide linked gamma subunits. Clone 9E1 specifically recognizes the extracellular D1 domain of the Fc epsilon R1 alpha chain. Fc epsilon R1 is primarily expressed on mast cells and basophils but expression of Fc epsilon R1 has also been reported on monocytes, Langerhans cells and dendritic cells from patients with atopic diseases.
Especies diana Human
Contenido y almacenamiento Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgG2b
Mostrar más Mostrar menos
For Research Use Only