Learn More
Fasudil monohydrochloride, 99+%, Thermo Scientific™
Marca: Thermo Scientific Alfa Aesar J60751.MD
| Cantidad | 250mg |
|---|
Descripción
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
SolubilitySoluble in water and dimethyl sulfoxide. Insoluble in ethanol.
Notes
Incompatible with acids, bases and oxidizing agents.
| 105628-07-7 | |
| 327.827 | |
| LFVPBERIVUNMGV-UHFFFAOYSA-N | |
| 163751 | |
| C1CNCCN(C1)S(=O)(=O)C2=CC=CC3=C2C=CN=C3.Cl |
| C14H18ClN3O2S | |
| MFCD00943198 | |
| fasudil hydrochloride, fasudil hcl, eril, at 877 hydrochloride, fasudil monohydrochloride, ha 1077 hydrochloride, unii-sq04n8s7br, 5-1,4-diazepan-1-yl sulfonyl isoquinoline hydrochloride, sq04n8s7br | |
| 5-(1,4-diazepan-1-ylsulfonyl)isoquinoline;hydrochloride |
Especificaciones
| Fasudil monohydrochloride | |
| C14H18ClN3O2S | |
| 250mg | |
| fasudil hydrochloride, fasudil hcl, eril, at 877 hydrochloride, fasudil monohydrochloride, ha 1077 hydrochloride, unii-sq04n8s7br, 5-1,4-diazepan-1-yl sulfonyl isoquinoline hydrochloride, sq04n8s7br | |
| LFVPBERIVUNMGV-UHFFFAOYSA-N | |
| 5-(1,4-diazepan-1-ylsulfonyl)isoquinoline;hydrochloride | |
| 163751 | |
| ≥99% |
| 105628-07-7 | |
| MFCD00943198 | |
| 14,3942 | |
| Soluble in water and dimethyl sulfoxide. Insoluble in ethanol. | |
| C1CNCCN(C1)S(=O)(=O)C2=CC=CC3=C2C=CN=C3.Cl | |
| 327.827 | |
| 327.83 |