Learn More
FANCD2 Antibody (1290D) - Azide and BSA Free, Novus Biologicals™
Código de producto. 18275698 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mg
Tamaño de la unidad:
0.10 miligramo
18275698 0.1 mg 0.10 miligramo
1 options
Código de producto. 18275698 Proveedor Novus Biologicals N.º de proveedor NBP280729
Rabbit Monoclonal Antibody
FANCD2 Monoclonal specifically detects FANCD2 in Human samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Flow (Intracellular), CyTOF-ready, Knockout Validated.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno FANCD2
Aplicaciones Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF, Immunoassay
Clasificación Monoclonal
Clon 1290D
Concentración 1 mg/mL
Conjugado Unconjugated
Dilución Western Blot : 0.1 - 0.5 μg/mL, Simple Western : 1 μg/mL, Flow Cytometry 1 μg/mL, Immunohistochemistry : 5-10 μg/mL, Immunohistochemistry-Paraffin : 5-10 μg/mL, Flow (Intracellular), CyTOF-ready, Knockout Validated
Formulación PBS
Alias de gen DKFZp762A223, FA4, FACD, FAD, FAD2, FA-D2, FADFAD2, FANCD, Fanconi anemia complementation group D2, Fanconi anemia group D2 protein, Fanconi anemia, complementation group D2, FLJ23826, FPN1, HFE4, IREG1, Protein FACD2, SLC11A3
Símbolos de los genes FANCD2
Especie del huésped Rabbit
Inmunógeno This FANCD2 Antibody (1290D) - Azide and BSA Free was developed against human FANCD2 fusion protein (N-terminal fragment). [Swiss-Prot #Q9BXW9]
Peso molecular del antígeno 164.1 kDa
Método de purificación Protein A or G purified from cell culture supernatant
Cantidad 0.1 mg
Estado normativo RUO
Disciplina de investigación Breast Cancer, Cancer, DNA Double Strand Break Repair, DNA Repair, Epigenetics, Genes Sensitive to DNA Damaging Agents
Primario o secundario Primary
ID de gen (Entrez) 2177
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgG