Learn More
Falnidamol, MedChemExpress
Código de producto. 16473808
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
50 mg
Tamaño de la unidad:
10 miligramos
5 miligramos
50 miligramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
16473808 10 mg 10 miligramos
16450187 50 mg 50 miligramos
16436537 5 mg 5 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 16473808 Proveedor Medchem Express N.º de proveedor HY10322
MedChemExpress Falnidamol (BIBX 1382) is an orally active, selective EGFR tyrosine kinase inhibitor with an IC50 of 3 nM. Falnidamol displays > 1000-fold lower potency against ErbB2 (IC50=3.4 μM) and a range of other related tyrosine kinases (IC50>10 μM). Falnidamol is a pyrimido-pyrimidine compound and has anti-cancer activity.
Falnidamol (BIBX 1382) is an orally active, selective EGFR tyrosine kinase inhibitor with an IC50 of 3 nM. Falnidamol displays > 1000-fold lower potency against ErbB2 (IC50=3.4 μM) and a range of other related tyrosine kinases (IC50>10 μM). Falnidamol is a pyrimido-pyrimidine compound and has anti-cancer activity.
Especificaciones
Nombre del producto químico o material Falnidamol
CAS 196612-93-8
Color Brownish Yellow
Notas de grado de pureza Research
Fórmula molecular C18H19ClFN7
Cantidad 10 mg
Sinónimo BIBX 1382
Información de solubilidad DMSO : 31.25 mg/mL (80.57 mM; ultrasonic and warming and heat to 60°C)
SMILES ClC1=CC(NC2=NC=NC3=CN=C(NC(CC4)CCN4C)N=C23)=CC=C1F
Peso molecular (g/mol) 387.84
Formula Weight (peso de la fórmula) 387.84
Porcentaje de pureza 97.03%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Cancer-Kinase/protease