missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Exosome component 1 Antibody [DyLight 405], Novus Biologicals Biologicals™
Tienda Bio Techne ProductosDescripción
Exosome component 1 Polyclonal antibody specifically detects Exosome component 1 in Human samples. It is validated for ELISA,Western Blot
Especificaciones
Especificaciones
| Antígeno | Exosome component 1 |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Polyclonal |
| Conjugado | DyLight 405 |
| Formulación | 50mM Sodium Borate |
| Alias de gen | CGI-108,3'-5' exoribonuclease CSL4 homolog, CSL4 exosomal core protein homolog, Csl4p, CSL4p13, exosomal core protein CSL4, exosome component 1hCsl4p, homolog of yeast exosomal core protein CSL4, SKI4, Ski4p |
| Especie del huésped | Rabbit |
| Inmunógeno | Recombinant fusion protein containing a sequence corresponding to amino acids 1-195 of human Exosome component 1 (NP_057130.1).,, Sequence:, MAPPVRYCIPGERLCNLEEGSPGSGTYTRHGYIFSSLAGCLMKSSENGALPVVSVVRETESQLLPDVGAIVTCKVSSINSRFAKVHILYVGSMPLKNSFRGTIRKEDVRATEKDKVEIYKSFRPGDIVLAKVISLGDAQSNYLLTTAENELGVVVAHSESGIQMVPISWCEMQCPKTHTKEFRKVARVQPEFLQT |
| Método de purificación | Affinity purified |
| Cantidad | 0.1 mL |
| Mostrar más |
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?