missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
ESX1 Polyclonal antibody specifically detects ESX1 in Human,Mouse samples. It is validated for ELISA,Immunoprecipitation,Western Blot
Especificaciones
Especificaciones
| Antígeno | ESX1 |
| Aplicaciones | ELISA, Immunoprecipitation, Western Blot |
| Clasificación | Polyclonal |
| Conjugado | DyLight 755 |
| Formulación | 50mM Sodium Borate |
| Alias de gen | ESX homeobox 1, ESX1LESX1R, ESX1-related protein, ESXR1homeobox protein ESX1, Extraembryonic, spermatogenesis, homeobox 1, extraembryonic, spermatogenesis, homeobox 1 homolog, extraembryonic, spermatogenesis, homeobox 1 homolog (mouse) |
| Especie del huésped | Rabbit |
| Inmunógeno | ESX homeobox 1. (Uniprot# Q8N693) |
| Método de purificación | Immunogen affinity purified |
| Cantidad | 0.1 mL |
| Mostrar más |
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?