Learn More
Berkshire International Ergonomic 16 in. Cleanroom Flat mop holder with clips
Tienda Berkshire ProductosDescripción
- Easy insertion of the holder into the pouches of the mop thanks to the V shape
- Ergonomic working hand-switch opening of the holder and mop discarding while standing upright
- No touching of soiled mops when discarding
- Optimum contact of the holder for very good particle removal
- Light weight makes it ideal for cleaning walls and ceilings
- Extremely maneuverable and smooth-running holder
- Autoclavable
- Detachable wiper clips allow the use of disposable mop wipers in addition to the foam or nonwoven mop covers but the clips are not autoclavable
Especificaciones
Especificaciones
| Tipo de producto | Cleanroom Mop |
| Esterilidad | Non-sterile |
| Esterilizables en autoclave | Autoclavable |
| Desechables | Reusable |
| Color-coded | No |
| Material | Polypropylene |