Learn More
Epirubicin hydrochloride, Thermo Scientific™
Cell-permeable anthracycline anti-tumor antibiotic
Marca: Thermo Scientific Alfa Aesar J60411.MA
| Cantidad | 10mg |
|---|
Descripción
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
| 56390-09-1 | |
| 579.983 | |
| MWWSFMDVAYGXBV-DPZLWMTJSA-N | |
| 123134690 | |
| CC1C(C(CC(O1)OC2CC(CC3=C(C4=C(C(=C23)O)C(=O)C5=C(C4=O)C=CC=C5OC)O)(C(=O)CO)O)N)O.Cl |
| C27H30ClNO11 | |
| MFCD00941448 | |
| Epiadriamycin | |
| (7S,9S)-7-[(2R,4R,5R)-4-amino-5-hydroxy-6-methyloxan-2-yl]oxy-6,9,11-trihydroxy-9-(2-hydroxyacetyl)-4-methoxy-8,10-dihydro-7H-tetracene-5,12-dione;hydrochloride |
Especificaciones
| Epirubicin hydrochloride | |
| C27H30ClNO11 | |
| 10mg | |
| Epiadriamycin | |
| MWWSFMDVAYGXBV-DPZLWMTJSA-N | |
| (7S,9S)-7-[(2R,4R,5R)-4-amino-5-hydroxy-6-methyloxan-2-yl]oxy-6,9,11-trihydroxy-9-(2-hydroxyacetyl)-4-methoxy-8,10-dihydro-7H-tetracene-5,12-dione;hydrochloride | |
| 123134690 |
| 56390-09-1 | |
| MFCD00941448 | |
| 14,3623 | |
| Soluble in DMSO to 100mM, or in ethanol to 10mM | |
| CC1C(C(CC(O1)OC2CC(CC3=C(C4=C(C(=C23)O)C(=O)C5=C(C4=O)C=CC=C5OC)O)(C(=O)CO)O)N)O.Cl | |
| 579.983 | |
| 579.98 |