Learn More
eIF2A, p Ser51 Antibody, Novus Biologicals™
Código de producto. 18211195 Tienda Bio Techne Productos
Cambiar vista
Click to view available options
Cantidad:
0.025 mg
0.1 mg
Tamaño de la unidad:
0.05 miligramo
0.10 miligramo
25 microgramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
18211195 0.025 mg 25 microgramos
18767253 - 0.05 miligramo
18527790 0.1 mg 0.10 miligramo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 18211195 Proveedor Novus Biologicals N.º de proveedor NB100818960.025MG
Rabbit Polyclonal Antibody has been used in 2 publications
eIF2A Polyclonal specifically detects eIF2A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno eIF2A (p Ser51)
Aplicaciones Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Clasificación Polyclonal
Concentración 1.0 mg/mL
Conjugado Unconjugated
Dilución Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:100-1:200, Immunohistochemistry-Paraffin 1:50-1:100
Formulación phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, and 50% glycerol. with 0.02% Sodium Azide
N.º de referencia del gen P05198
Alias de gen EIF-2, EIF-2A, EIF2AEIF2, EIF-2alpha, eukaryotic translation initiation factor 2, subunit 1 alpha, 35kDa, subunit 1 (alpha, 35kD )
Símbolos de los genes EIF2A
Especie del huésped Rabbit
Inmunógeno The antiserum was produced against synthesized phosphopeptide derived from human eIF2. around the phosphorylation site of serine 51 (E-L-SP-R-R).
Peso molecular del antígeno 38 kDa
Método de purificación Affinity Purified
Cantidad 0.025 mg
Estado normativo RUO
Disciplina de investigación DNA Repair, Phospho Specific, Translation Control, Unfolded Protein Response
Primario o secundario Primary
ID de gen (Entrez) 83939
Especificidad de la prueba eIF2. (specific to Phospho-Ser51) detects endogenous levels of eIF2. only when phosphorylated at serine 51.
Especies diana Mouse, Rat, Human
Contenido y almacenamiento Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Mostrar más Mostrar menos
For Research Use Only