missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
EGF Monoclonal specifically detects EGF in Human samples. It is validated for ELISA.
Specifications
Specifications
| Antígeno | EGF |
| Aplicaciones | ELISA |
| Clasificación | Monoclonal |
| Clon | 008 |
| Conjugado | Alexa Fluor 750 |
| Dilución | ELISA |
| Formulación | 50mM Sodium Borate |
| Alias de gen | beta-urogastrone, epidermal growth factor, epidermal growth factor (beta-urogastrone), hEGF, HOMG4, pro-epidermal growth factor, URG |
| Especie del huésped | Rabbit |
| Inmunógeno | This antibody was obtained from a rabbit immunized with purified, recombinant Human EGF (NP_001954.2; Asn 971-Arg 1023) . |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?