Learn More
Diacerein, Thermo Scientific™
Interleukin-1 inhibitor
Marca: Thermo Scientific Alfa Aesar J63775.03
| Cantidad | 1g |
|---|
Descripción
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
| 13739-02-1 | |
| 368.30 | |
| TYNLGDBUJLVSMA-UHFFFAOYSA-N | |
| 26248 | |
| CC(=O)OC1=CC=CC2=C1C(=O)C1=C(OC(C)=O)C=C(C=C1C2=O)C(O)=O |
| C19H12O8 | |
| MFCD00468030 | |
| diacerein, diacetylrhein, diacerhein, artrodar, fisiodar, 1,8-diacetoxy-3-carboxyanthraquinone, 4,5-diacetylrhein, diacerin, diacerein inn, diacereine french | |
| 4,5-bis(acetyloxy)-9,10-dioxo-9,10-dihydroanthracene-2-carboxylic acid |
Especificaciones
| Diacerein | |
| C19H12O8 | |
| 1g | |
| diacerein, diacetylrhein, diacerhein, artrodar, fisiodar, 1,8-diacetoxy-3-carboxyanthraquinone, 4,5-diacetylrhein, diacerin, diacerein inn, diacereine french | |
| CC(=O)OC1=CC=CC2=C1C(=O)C1=C(OC(C)=O)C=C(C=C1C2=O)C(O)=O | |
| 368.30 | |
| 368.29 |
| 13739-02-1 | |
| MFCD00468030 | |
| 14,2960 | |
| TYNLGDBUJLVSMA-UHFFFAOYSA-N | |
| 4,5-bis(acetyloxy)-9,10-dioxo-9,10-dihydroanthracene-2-carboxylic acid | |
| 26248 |