missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Death Ligand Signal Enhancer Antibody, Alexa Fluor™ 700, Novus Biologicals™
Tienda Bio Techne ProductosDescription
Death Ligand Signal Enhancer Polyclonal antibody specifically detects Death Ligand Signal Enhancer in Human, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
Specifications
| Antígeno | Death Ligand Signal Enhancer |
| Aplicaciones | Western Blot, ELISA, Immunofluorescence |
| Clasificación | Polyclonal |
| Conjugado | Alexa Fluor 700 |
| Formulación | 50mM Sodium Borate |
| Alias de gen | DAP3 Binding Cell Death Enhancer 1, DAP3-Binding Cell Death Enhancer 1, DAP3-Binding Cell Death Enhancer 1, Long Form, Death Ligand Signal Enhancer, DELE, DELE1, DELE1(L), hypothetical protein LOC9812, KIAA0141 |
| Especie del huésped | Rabbit |
| Inmunógeno | Death Ligand Signal Enhancer antibody was raised against an 18 amino acid synthetic peptide near the amino terminus of human Death Ligand Signal Enhancer. The immunogen is located within the first 50 amino acids of Death Ligand Signal Enhancer. Amino Acid Squence: RVTPKSTSPDGPQTTSST |
| Método de purificación | Peptide affinity purified |
| Cantidad | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?