Learn More
Eurofins DiscoverX DDR1 Protein, Active
Descripción
Tyrosine kinase that functions as cell surface receptor for fibrillar collagen and regulates cell attachment to the extracellular matrix, remodeling of the extracellular matrix, cell migration, differentiation, survival and cell proliferation. Collagen binding triggers a signaling pathway that involves SRC and leads to the activation of MAP kinases. Regulates remodeling of the extracellular matrix by up-regulation of the matrix metalloproteinases MMP2, MMP7 and MMP9, and thereby facilitates cell migration and wound healing. Required for normal blastocyst implantation during pregnancy, for normal mammary gland differentiation and normal lactation. Required for normal ear morphology and normal hearing (By similarity). Promotes smooth muscle cell migration, and thereby contributes to arterial wound healing. Also plays a role in tumor cell invasion. Phosphorylates PTPN11.
GenBank U48705
Especificaciones
Especificaciones
| Formulación | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 20 mM β-glycerophosphate, 0.25 mM sodium orthovanadate, 10 mM NaF, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
| Peso molecular | 50 kDa |
| Cantidad | 250 μg |
| Especie | Human |
| Recombinante | Recombinant |
| Etiqueta de proteína | N-terminal 6His |
| Sistema de expresión | Recombinant enzyme expressed in Sf21 insect cells |
| Forma de proteína | Truncated |
| Grado de pureza o calidad | ≥62% |
| Expresión | DDR1, activated |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.