Learn More
DDIT4L Antibody (DDIT-03), Janelia Fluor™ 646, Novus Biologicals™
Código de producto. 18275733 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
18275733 0.1 mL 0.10 ml
1 options
Código de producto. 18275733 Proveedor Novus Biologicals N.º de proveedor NBP262218JF646
Mouse Monoclonal Antibody
DDIT4L Monoclonal specifically detects DDIT4L in Human samples. It is validated for Western Blot, Flow Cytometry.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno DDIT4L
Aplicaciones Western Blot, Flow Cytometry
Clasificación Monoclonal
Clon DDIT-03
Conjugado Janelia Fluor 646
Dilución Western Blot, Flow Cytometry
Formulación 50mM Sodium Borate with 0.05% Sodium Azide
Alias de gen DNA damage-inducible transcript 4-like protein, DNA-damage-inducible transcript 4-like, HIF-1 responsive protein RTP801-like, homolog of mouse SMHS1, Protein regulated in development and DNA damage response 2, REDD-2, REDD2Rtp801L, regulated in development and DNA damage response 2, RTP801L
Símbolos de los genes DDIT4L
Especie del huésped Mouse
Inmunógeno N-terminal recombinant fragment of human DDIT4L (amino acids 2-98)
Método de purificación Protein A purified
Cantidad 0.1 mL
Disciplina de investigación Cancer, mTOR Pathway
Primario o secundario Primary
ID de gen (Entrez) 115265
Especificidad de la prueba The antibody MEM-18 reacts with CD14, a 53-55 kDa GPI (glycosylphosphatidylinositol)-linked membrane glycoprotein expressed on monocytes, macrophages and weakly on granulocytes; also expressed by most tissue macrophages. In human, the epitope recognized by MEM-18 is located between amino acids 57-64.
Especies diana Human
Contenido y almacenamiento Store at 4°C in the dark.
Isotype IgG1