Learn More
DC260126, MedChemExpress
Código de producto. 16474716
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
25 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
25 miligramos
5 miligramos
50 miligramos
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
16474716 100 mg 100 miligramos
16450027 50 mg 50 miligramos
16432806 25 mg 25 miligramos
16426367 5 mg 5 miligramos
16403638 10 mg 10 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Código de producto. 16474716 Proveedor Medchem Express N.º de proveedor HY101906
MedChemExpress DC260126 is a potent antagonist of GPR40 (FFAR1). DC260126 dose-dependently inhibits GPR40-mediated Ca2+ elevations stimulated by linoleic acid, oleic acid, palmitoleic acid and lauric acid (IC50: 6.28, 5.96, 7.07, 4.58 μM, respectively). DC260126 could protect MIN6 β cells from palmitate-induced ER stress and apoptosis.
DC260126 is a potent antagonist of GPR40 (FFAR1). DC260126 dose-dependently inhibits GPR40-mediated Ca2+ elevations stimulated by linoleic acid, oleic acid, palmitoleic acid and lauric acid (IC50: 6.28, 5.96, 7.07, 4.58 μM, respectively). DC260126 could protect MIN6 β cells from palmitate-induced ER stress and apoptosis.
Especificaciones
Nombre del producto químico o material DC260126
CAS 346692-04-4
Color Blanco
Notas de grado de pureza Research
Fórmula molecular C16H18FNO2S
Cantidad 100 mg
Información de solubilidad DMSO : ≥ 100 mg/mL (325.33 mM)
SMILES O=S(C1=CC=C(F)C=C1)(NC2=CC=C(CCCC)C=C2)=O
Peso molecular (g/mol) 307.38
Formula Weight (peso de la fórmula) 307.38
Porcentaje de pureza 99.74%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Metabolism-sugar/lipid metabolism