Learn More
DAPT, MedChemExpress
Código de producto. 16491667
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
50 mg
Tamaño de la unidad:
10 miligramos
5 miligramos
50 miligramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
16491667 50 mg 50 miligramos
16478207 5 mg 5 miligramos
16435498 10 mg 10 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 16491667 Proveedor Medchem Express N.º de proveedor HY13027
MedChemExpress DAPT (GSI-IX) is a potent and orally active γ-secretase inhibitor with IC50s of 115 nM and 200 nM for total amyloid-β (Aβ) and Aβ42, respectively.
DAPT inhibits the activation of Notch 1 signaling and induces cell differentiation. DAPT also induces autophagy and apoptosis. DAPT has neuroprotection activity and has the potential for autoimmune and lymphoproliferative diseases, degenerative disease and cancers treatment.
Especificaciones
Nombre del producto químico o material DAPT
CAS 208255-80-5
Color Blanco
Notas de grado de pureza Research
Fórmula molecular C23H26F2N2O4
Cantidad 50 mg
Sinónimo GSI-IX
Información de solubilidad DMSO : 62.5 mg/mL (144.52 mM; Need ultrasonic) ∣Ethanol : 10 mg/mL (23.12 mM; Need ultrasonic)
SMILES FC1=CC(F)=CC(CC(N[C@H](C(N[C@H](C(OC(C)(C)C)=O)C2=CC=CC=C2)=O)C)=O)=C1
Peso molecular (g/mol) 432.46
Formula Weight (peso de la fórmula) 432.46
Porcentaje de pureza 99.62%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Cancer-programmed cell death