Learn More
CYB5R3 Antibody (S01-9F3), Novus Biologicals™
Código de producto. 18605174 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
100 μg
50 μg
Tamaño de la unidad:
100 microlitros
50 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
18605174 50 μg 50 microlitros
18604105 100 μg 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 18605174 Proveedor Novus Biologicals N.º de proveedor NBP31961450ul
Rabbit Monoclonal Antibody
CYB5R3 Monoclonal antibody specifically detects CYB5R3 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno CYB5R3
Aplicaciones Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Clasificación Monoclonal
Clon S01-9F3
Concentración 0.3 mg/mL
Conjugado Unconjugated
Dilución Western Blot, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin
Formulación 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Alias de gen B5RNADH-cytochrome b5 reductase 3, Cytochrome b5 reductase, cytochrome b5 reductase 3, DIA1NADH-cytochrome b5 reductase 3 membrane-bound form, diaphorase (NADH) (cytochrome b-5 reductase), Diaphorase-1, EC 1.6.2.2, NADH-cytochrome b5 reductase 3 soluble form
Especie del huésped Rabbit
Inmunógeno A synthetic peptide of human CYB5R3 (Uniprot # P00387)
Método de purificación Affinity purified
Cantidad 50 μg
Estado normativo RUO
Disciplina de investigación Cancer, Lipid and Metabolism
Primario o secundario Primary
ID de gen (Entrez) 1727
Especies diana Human, Rat
Contenido y almacenamiento Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgG