Learn More
Crizotinib hydrochloride, MedChemExpress
Código de producto. 16471926
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
10 mg
50 mg
100 mg
200 mg
500 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
200 miligramos
50 miligramos
500 miligramos
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
16471926 500 mg 500 miligramos
16471326 200 mg 200 miligramos
16448128 10 mg 10 miligramos
16447886 100 mg 100 miligramos
16404047 50 mg 50 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Código de producto. 16471926 Proveedor Medchem Express N.º de proveedor HY50878A
MedChemExpress Crizotinib hydrochloride (PF-02341066 hydrochloride) is an orally bioavailable, selective, and ATP-competitive dual ALK and c-Met inhibitor with IC50s of 20 and 8 nM, respectively.
Crizotinib hydrochloride (PF-02341066 hydrochloride) inhibits tyrosine phosphorylation of NPM-ALK and tyrosine phosphorylation of c-Met with IC50s of 24 and 11 nM in cell-based assays, respectively. It is also a ROS proto-oncogene 1 (ROS1) inhibitor. Crizotinib hydrochloride (PF-02341066 hydrochloride) has effective tumor growth inhibition.
Especificaciones
Nombre del producto químico o material Crizotinib hydrochloride
CAS 1415560-69-8
Color Earth Yellow
Notas de grado de pureza Research
Fórmula molecular C21H23Cl3FN5O
Cantidad 500 mg
Sinónimo PF-02341066 hydrochloride
Información de solubilidad H2O : 50 mg/mL (102.71 mM; Need ultrasonic) ∣DMSO : ≥ 4.9 mg/mL (10.07 mM)
SMILES ClC1=C(F)C=CC(Cl)=C1[C@H](OC2=CC(C3=CN(N=C3)C4CCNCC4)=CN=C2N)C.[H]Cl
Peso molecular (g/mol) 486.8
Formula Weight (peso de la fórmula) 486.8
Porcentaje de pureza 99.13%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Cancer-Kinase/protease