missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
COX8A Polyclonal antibody specifically detects COX8A in Human,Mouse samples. It is validated for ELISA,Immunohistochemistry,Western Blot
Especificaciones
Especificaciones
| Antígeno | COX8A |
| Aplicaciones | ELISA, Immunohistochemistry, Western Blot |
| Clasificación | Polyclonal |
| Conjugado | DyLight 488 |
| Formulación | 50mM Sodium Borate |
| Alias de gen | COX, COX8-2, COX8cytochrome c oxidase subunit VIII, COX8Lcytochrome c oxidase subunit 8A (ubiquitous), Cytochrome c oxidase polypeptide VIII-liver/heart, Cytochrome c oxidase subunit 8-2, cytochrome c oxidase subunit 8A, mitochondrial, cytochrome c oxidase subunit VIIIA (ubiquitous), VIII, VIII-L |
| Especie del huésped | Rabbit |
| Inmunógeno | cytochrome c oxidase subunit 8A(ubiquitous). (Uniprot# P10176) |
| Método de purificación | Immunogen Affinity-purified |
| Cantidad | 0.1 mL |
| Mostrar más |
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?