missing translation for 'onlineSavingsMsg'
Learn More

COG1 Antibody [mFluor Violet 450 SE], Novus Biologicals™

Product Code. 30608217 Shop All Bio Techne Products
Change view
Click to view available options
Cantidad:
0.1 mL
Unit Size:
0.10 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
30608217 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30608217 Supplier Novus Biologicals Supplier No. NBP347565MFV450

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

COG1 Polyclonal antibody specifically detects COG1 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antígeno COG1
Aplicaciones ELISA, Immunohistochemistry, Western Blot
Clasificación Polyclonal
Conjugado mFluor Violet 450 SE
Formulación 50mM Sodium Borate
Alias de gen COG complex subunit 1, component of oligomeric golgi complex 1CDG2G, conserved oligomeric Golgi complex subunit 1, KIAA1381DKFZp762L1710, LDLBconserved oligomeric Golgi complex protein 1, low density lipoprotein receptor defect B complementing
Especie del huésped Rabbit
Inmunógeno component of oligomeric golgi complex 1. (Uniprot# Q8WTW3)
Método de purificación Immunogen Affinity-purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Signal Transduction
Primario o secundario Primary
ID de gen (Entrez) 9382
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.