Learn More
Cobalt bis(trifluoromethylsulfonyl)imide, Thermo Scientific™
Marca: Thermo Scientific Alfa Aesar 047053.03
| Cantidad | 1g |
|---|
Descripción
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
| 207861-61-8 | |
| 619.204 | |
| HRURXKIZWNSHQB-UHFFFAOYSA-N | |
| 131875824 | |
| C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Co+2] |
| C4CoF12N2O8S4 | |
| MFCD23380169 | |
| cobalt bis-trifluoromethanesulfonimide | |
| bis(trifluoromethylsulfonyl)azanide;cobalt(2+) |
Especificaciones
| Cobalt bis(trifluoromethylsulfonyl)imide | |
| 1g | |
| [(CF3SO2)2N]2Co | |
| UN3261 | |
| It is soluble in water. | |
| C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Co+2] | |
| 619.204 | |
| 619.23 |
| 207861-61-8 | |
| C4CoF12N2O8S4 | |
| MFCD23380169 | |
| cobalt bis-trifluoromethanesulfonimide | |
| HRURXKIZWNSHQB-UHFFFAOYSA-N | |
| bis(trifluoromethylsulfonyl)azanide;cobalt(2+) | |
| 131875824 | |
| Hygroscopic |